Anti-CCR10 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H55]
Catalog # USBIC2100-16A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 10
- Antigen symbol:CCR10
- Clonality:Monoclonal
- Clone:7H55
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Rat,Mouse
- Form:liquid
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,01 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CCR10 (chemokine (C-C motif) receptor 10)
Clonality: Monoclonal
Clone: 7H55
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Rat