Anti-Bombesin Rabbit polyclonal antibody unconjugated
USBIB2550-51
- Antibody type:Primary
- Antigen name:Bombesin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig
- Format:Serum
- Form:lyophilized powder
- Sequence:pEQRLGNQWAVGHLM-NH2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500
Type: Primary
Antigen: Bombesin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig