Anti-Bombesin Rabbit polyclonal antibody unconjugated
Catalog # USBIB2550-05
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Bombesin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken,Human
- Form:liquid
- Storage buffer:PBS, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:250 µl
- Pk:250 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:30-1:300; Immunochemistry: 1:400-1:2000
Type: Primary
Antigen: Bombesin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human