Anti-D-Aspartic Acid Rat polyclonal antibody unconjugated
Catalog # USBIA3879-30B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:D-Aspartic Acid
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by protein carrier (BSA) affinity column, and ammonium sulphate precipitation
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: D-Aspartic Acid
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: