Order Entry
France
ContactUsLinkComponent
Anti-ARAP1 Goat Polyclonal Antibody
Anti-ARAP1 Goat Polyclonal Antibody
Catalog # USBIA3309-02
Supplier:  US Biological
Anti-ARAP1 Goat Polyclonal Antibody
Catalog # USBIA3309-02
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
  • Antigen symbol:
    ARAP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
  • Sequence:
    C-AFTADPLSLLRNV
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: ARAP1 (ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat