Order Entry
France
ContactUsLinkComponent
 
Anti-APP Mouse monoclonal antibody unconjugated [clone: 9L22]
  USBIA2275-59E
Anti-APP Mouse monoclonal antibody unconjugated [clone: 9L22]
  USBIA2275-59E

 

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid Beta (A4) Precursor Protein
  • Antigen symbol:
    APP
  • Clonality:
    Monoclonal
  • Clone:
    9L22
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Antigen synonyms:
    ABETA|beta Amyloid
  • Storage buffer:
    2X PBS, PEG, sucrose, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by thiophilic adsorption and size exclusion chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: 9L22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human