Anti-APP Mouse monoclonal antibody unconjugated [clone: 12K249]
Catalog # USBIA2275-59C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Amyloid Beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Monoclonal
- Clone:12K249
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:N-terminal
- Form:lyophilized powder
- Antigen synonyms:ABETA|beta Amyloid
- Storage buffer:0,01M PBS, pH 7,2. ddH2O to 1mg/ml.
- Sequence:DAEFRHDS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: 12K249
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human