Anti-Amphetamine Mouse Monoclonal Antibody [clone: 10B152]
Catalog # USBIA1510-02
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Amphetamine
- Clonality:Monoclonal
- Clone:10B152
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Amphetamine
Clonality: monoclonal
Clone: 10B152
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: