Order Entry
France
ContactUsLinkComponent
Anti-IFNB1 Rabbit Polyclonal Antibody
Anti-IFNB1 Rabbit Polyclonal Antibody
Catalog # USBI146171
Supplier:  US Biological
Anti-IFNB1 Rabbit Polyclonal Antibody
Catalog # USBI146171
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interferon Beta 1, fibroblast
  • Antigen symbol:
    IFNB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 5 ?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: IFNB1 (interferon, beta 1, fibroblast)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse