Anti-ERBB2IP Sheep Polyclonal Antibody
Catalog # USBI145458
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:erbb2 interacting Protein
- Antigen symbol:ERBB2IP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.2?g/ml; Immunochemistry: 515 ?g/ml
Type: Primary
Antigen: ERBB2IP (erbb2 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: