Anti-DDB1 Mouse Monoclonal Antibody [clone: 13k42]
Catalog # USBI143169
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:damage-specific DNA binding Protein 1, 127kDa
- Antigen symbol:DDB1
- Clonality:Monoclonal
- Clone:13k42
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:ascites fluid.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
- Recommended dilution: Western Blot: 1:1500
Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: monoclonal
Clone: 13k42
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse, Rat