Anti-HNRNPD Rabbit Monoclonal Antibody [clone: 13G38]
Catalog # USBI137446
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding Protein 1, 37kDa)
- Antigen symbol:HNRNPD
- Clonality:Monoclonal
- Clone:13G38
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol and <0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: HNRNPD (heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa))
Clonality: monoclonal
Clone: 13G38
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human