Anti-FBXO25 Mouse Monoclonal Antibody [clone: 2F1]
USBI126677
- Antibody type:Primary
- Antigen name:F-box protein 25
- Antigen symbol:FBXO25
- Clonality:Monoclonal
- Clone:2F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MPFLGQDWRSPGWSWIKTEDGWKRCESCSQKLERENNHCNISHSIILNSEDGEIFNNEEHEYASKKRKKDHFRNDTNTQSFYREKWIYVHKESTKERHGYCTLGEAFNRLDFSSAIQDIRRFNYVVKLLQLIAKSQLTSLSGVAQKNYFNILDKIVQKVLDDHHNPRLIKDLLQDLSSTLCILIRGVGKSVLVGNINIWICRLETILAWQQQLQDLQMTKQVNNGLTLSDLPLHMLNNILYRFSDGWDIITLGQV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: FBXO25 (F-box protein 25)
Clonality: monoclonal
Clone: 2F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human