Anti-DVL3 Rabbit Polyclonal Antibody
USBI126084
- Antibody type:Primary
- Antigen name:Dishevelled segment polarity protein 3
- Antigen symbol:DVL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: DVL3 (dishevelled segment polarity protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human