Anti-DOCK2 Mouse Monoclonal Antibody [clone: 2F4]
Catalog # USBI125984
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 2
- Antigen symbol:DOCK2
- Clonality:Monoclonal
- Clone:2F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MCVCAEDGKTLPVAVPIEDMQRIHLRFMFRHRSSLESKDKGEKNFAMSYVKLMKEDGTTLHDGFHDLVVLKGDSKKMEDASAYLTLPSYRHHVENKGATLSRSSSSVGGLSVSSRDVFSISTLVCSTKLTQNVGLLGLLKWRMKPQLEESMRRLFESINNLMKSQYKTTILLQVAALKYIPSVLHDVEMVFDAKLLSQLLYEFYTCIPPVKLQKQKVQSMNEIVQSNLFKKQECRDILLPVITKELKELLEQKDDMQHQVLERKYCVELLNSILEVLSYQDAAFTYHHIQEIMVQLLRTVNRTVITMGRDHILIGACCNCGFPSSTPHSLIQWLWG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DOCK2 (dedicator of cytokinesis 2)
Clonality: monoclonal
Clone: 2F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human