Order Entry
France
ContactUsLinkComponent
Anti-DOCK1 Mouse Monoclonal Antibody [clone: 3D10]
Anti-DOCK1 Mouse Monoclonal Antibody [clone: 3D10]
Catalog # USBI125982
Supplier:  US Biological
Anti-DOCK1 Mouse Monoclonal Antibody [clone: 3D10]
Catalog # USBI125982
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 1
  • Antigen symbol:
    DOCK1
  • Clonality:
    Monoclonal
  • Clone:
    3D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    698 to 803
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    LETYIKKHFSATLAYTKLTKVLKNYVDGAEKPGVNEQLYKAMKALESIFKFIVRSRILFNQLYENKGEADFVESLLQLFRSINDMMSSMSDQTVRVKGAALKYLPT
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: DOCK1 (dedicator of cytokinesis 1)
Clonality: monoclonal
Clone: 3D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human