Order Entry
France
ContactUsLinkComponent
Anti-COPS8 Mouse Monoclonal Antibody [clone: 2G8]
Anti-COPS8 Mouse Monoclonal Antibody [clone: 2G8]
  USBI125230
Anti-COPS8 Mouse Monoclonal Antibody [clone: 2G8]
  USBI125230

 

  • Antibody type:
    Primary
  • Antigen name:
    COP9 signalosome subunit 8
  • Antigen symbol:
    COPS8
  • Clonality:
    Monoclonal
  • Clone:
    2G8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MPVAVMAESAFSFKKLLDQCENQELEAPGGIATPPVYGQLLALYLLHNDMNNARYLWKRIPPAIKSANSELGGIWSVGQRIWQRDFPGIYTTINAHQWSETVQPIMEALRDATRRRAFALVSQAYTSIIADDFAAFVGLPVEEAVKGILEQGWQADSTTRMVLPRKPVAGALDVSFNKFIPLSEPAPVPPIPNEQQLARLTDYVAFLEN
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: COPS8 (COP9 signalosome subunit 8)
Clonality: monoclonal
Clone: 2G8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human