Anti-AUH Mouse Monoclonal Antibody [clone: 2G12]
USBI123757
- Antibody type:Primary
- Antigen name:AU RNA binding Protein/enoyl-CoA hydratase
- Antigen symbol:AUH
- Clonality:Monoclonal
- Clone:2G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:44 to 135
- Storage buffer:PBS, pH 7,2.
- Sequence:RAGPAIWAQGWVPAAGGPAPKRGYSSEMKTEDELRVRHLEEENRGIVVLGINRAYGKNSLSKNLIKMLSKAVDALKSDKKVRTIIIRSEVPG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: AUH (AU RNA binding protein/enoyl-CoA hydratase)
Clonality: monoclonal
Clone: 2G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human