Anti-ANGPTL4 Rabbit Polyclonal Antibody
Catalog # USBI044529
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Angiopoietin-like 4
- Antigen symbol:Aminomethyltransferase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:ANGPTL4
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:2000-1:5000; Western Blot: 1:2000-1:5000
Type: Primary
Antigen: ANGPTL4 (angiopoietin-like 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: