Order Entry
France
ContactUsLinkComponent
 
Anti-TSC2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI043346-PE
Anti-TSC2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI043346-PE

 

  • Antibody type:
    Primary
  • Antigen name:
    tuberous sclerosis 2
  • Antigen symbol:
    TSC2
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Modification:
    phospho-Ser1343
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Dot Blot, Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Dot blot 1:500
Do not freeze!

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse