Anti-LIMS2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI037789-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:LIM and senescent cell antigen-like domains 2
- Antigen symbol:LIMS2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:295 to 325
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: LIMS2 (LIM and senescent cell antigen-like domains 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse