Order Entry
France
ContactUsLinkComponent
 
Anti-LIM2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI037780-PE
undefined
Anti-LIM2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI037780-PE

 

  • Antibody type:
    Primary
  • Antigen name:
    lens intrinsic membrane Protein 2, 19kDa
  • Antigen symbol:
    LIM2
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Amino acid number:
    109 to 137
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: LIM2 (lens intrinsic membrane protein 2, 19kDa)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human