Anti-IL12RB2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI036973-FITC
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Interleukin 12 Receptor Beta 2
- Symbole de l'antigène:IL12RB2
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Épitope:C-terminal
- Formulaire:liquid
- Nombre d'acides aminés:759 to 788
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Température de stockage:Cool
- Température de transport:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: IL12RB2 (interleukin 12 receptor, beta 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human