Anti-GLTSCR2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI036104-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glioma tumor suppressor candidate region 2
- Antigen symbol:GLTSCR2
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:429 to 458
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: GLTSCR2 (glioma tumor suppressor candidate region gene 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human