Anti-TRDMT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI034743-FITC
- Antibody type:Primary
- Antigen name:tRNA aspartic acid methyltransferase 1
- Antigen symbol:TRDMT1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Form:liquid
- Amino acid number:190 to 220
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50;
Do not freeze!
Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human