Anti-TRDMT1 Rabbit Polyclonal Antibody
Catalog # USBI034741
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tRNA aspartic acid methyltransferase 1
- Antigen symbol:TRDMT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:353 to 383
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse