Saisie de commande
France
ContactUsLinkComponent
 
Anti-IgG (H+L) Donkey Polyclonal Antibody (Texas Red®)
  ROCK805-7902
Anti-IgG (H+L) Donkey Polyclonal Antibody (Texas Red®)
  ROCK805-7902

 

  • Type d'anticorps:
    Secondaire
  • Symbole de l'antigène:
    IgG (H+L)
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Texas Red®
  • ELISA:
    Yes
  • Cytométrie en flux:
    Yes
  • Changement de gel:
    Yes
  • Hôte:
    Donkey
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Précipitation immunologique:
    Yes
  • Réactivité:
    Goat
  • Western blot:
    Yes
  • Adsorption croisée:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Formulaire:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Température de stockage:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Température de transport:
    Ice
  • Immunogène:
    Goat IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Taille:
    1 mg
  • Cdt:
    1 mg

 

 

Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat