Anti-IgM µ Goat Polyclonal Antibody (PE (Phycoerythrin))
Catalog # ROCK712-1807
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Antibody fragment:F(ab')2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:0,5 mg/ml by absorbance = 82,0 at 565 nm
- Shipping temperature:Ice
- Immunogen:Rat IgM mu heavy chain
- Purification:purified by immunoaffinity chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rat