Anti-IgG F(c) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK710-1222
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG F(c)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:Bovine, Horse, Human
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Antibody fragment:F(ab')2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Mouse IgG F(c) fragment
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Mouse