Anti-Biotin Goat Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ROCK700-102-098
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Biotin
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Multiple
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Antibody fragment:F(ab')2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Biotin conjugated to Keyhole Limpet Hemocyanin (KLH)
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.
Recommended Dilutions: Immunofluorescence: 1:1000-1:5000; Flow Cytometry: 1:500-1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan