Anti-IgM µ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
ROCK612-4207
- Antibody type:Secondary
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Rat IgM mu heavy chain
- Purification:purified by immunoaffinity chromatography
- Size:1,5 mg
- Pk:1.500 µG
Secondary Rabbit Anti-IgM (mu chain) Reacts with Rat
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: µ-chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat