Anti-RFA2 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-447
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen symbol:RFA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Yeast
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:855404
- Amino acid number:115 to 129
- Modification:phospho-Ser122
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,54 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to aa 115-129 (of 273) of yeast DNA Replication Factor A (RFA2). The RFA complex consists of a heterotrimer of
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity-purified antibody was tested for use in western blotting against RFA2 containing cell lysates and by ELISA against the immunizing peptide. Reactivity in other immunoassays is unknown.
Recommended Dilutions: ELISA: 1:8000 - 1:20000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: RFA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast