Anti-ULP1 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-492
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen symbol:ULP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Yeast
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:856087
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:5,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This purified antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast ULP-1 Protein.
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Size:500 µg
- Pk:500 µG
Specifications
About this item
This purified polyclonal antibody reacts with yeast ULP-1 by western blot and ELISA.
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: ULP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast