Order Entry
France
ContactUsLinkComponent
Anti-Mouse Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2D11.H5]
Anti-Mouse Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2D11.H5]
Catalog # ROCK110-4101
Anti-Mouse Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2D11.H5]
Catalog # ROCK110-4101

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mouse Serum
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
  • Storage temperature:
    Cool
  • Concentration:
    80 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse serum Proteins
  • Purification:
    delipidation and defibrination
  • Size:
    2 ml
  • Pk:
    2 mL

Specifications

About this item

Anti-MOUSE SERUM (RABBIT) Antibody is suitable for immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring high titer and specificity.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Mouse Serum
Clonality: Monoclonal
Clone: 2D11.H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse