Anti-CUL4A Rabbit Monoclonal Antibody (Unconjugated) [clone: 8A3]
Catalog # ROCK100-401-A04
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:cullin 4A
- Antigen symbol:CUL4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:8451
- Amino acid number:1 to 20
- Storage buffer:0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 1-20 of Human Cul4A (N-terminus) coupled to KLH.
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Specifications
About this item
This antibody reacts with human Cul4A by western blot and immunoprecipitation. The antibody immunoprecipitates Protein from cell lysates (using HeLa, U2OS and others). This antibody also co-immunoprecipitates at least 8 subunits of COP9, DDB1 and CAND1.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: CUL4A
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human