Order Entry
France
ContactUsLinkComponent
Anti-V5 Tag Mouse Monoclonal Antibody
Anti-V5 Tag Mouse Monoclonal Antibody
Supplier:  Genscript
Certificates
undefined
Anti-V5 Tag Mouse Monoclonal Antibody
Supplier:  Genscript

Specifications

  • Catalog No:
  • GENSA01724-100
  • GENSA01724-200
  • Antigen symbol:
  • V5
  • V5
  • Antigen name:
  • V5 tag
  • V5 tag
  • Antibody type:
  • Tag
  • Tag
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together