Anti-Ty1-tag Rabbit Polyclonal Antibody
Catalog # GENSA01004-40
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen symbol:Ty1-tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:40 µg
- Pk:40 µG
Specifications
About this item
Type: Tag
Antigen: Ty1-tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: