Anti-HSPA4L Goat Polyclonal Antibody
Catalog # EVBIEB08505
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 70kDa protein 4-like
- Antigen symbol:HSPA4L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HSPA4L (heat shock 70kDa protein 4-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human