Anti-CSMD3 Goat polyclonal antibody unconjugated
Catalog # EVBIEB08318
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:CUB and Sushi multiple domains 3
- Antigen symbol:CSMD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CSMD3 (CUB and Sushi multiple domains 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat