Anti-TMPRSS4 Goat Polyclonal Antibody
Catalog # EVBIEB08297
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:transmembrane protease, serine 4
- Antigen symbol:TMPRSS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TMPRSS4 (transmembrane protease, serine 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse