Anti-SLC2A8 Goat Polyclonal Antibody
Catalog # EVBIEB07322
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 2 (facilitated glucose transporter), member 8
- Antigen symbol:Solute carrier family 2, member 8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SLC2A8 (solute carrier family 2 (facilitated glucose transporter), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human