Anti-GLTSCR2 Goat Polyclonal Antibody
EVBIEB06549
- Antibody type:Primary
- Antigen name:Glioma tumor suppressor candidate region 2
- Antigen symbol:GLTSCR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: GLTSCR2 (glioma tumor suppressor candidate region gene 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: