Anti-FANCG Goat Polyclonal Antibody
Catalog # EVBIEB06113
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Fanconi anemia, complementation group G
- Antigen symbol:FANCG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Dog
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: FANCG (Fanconi anemia, complementation group G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Pig