Anti-LIF Rat Monoclonal Antibody [clone: 39N7D10]
Catalog # NOVUNBP2-27406
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:leukemia inhibitory factor
- Antigen symbol:LIF
- Clonality:Monoclonal
- Clone:39N7D10
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:3976
- Antigen synonyms:MLPLI|DIA|CDF|HILDA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: LIF (leukemia inhibitory factor)
Clonality: monoclonal
Clone: 39N7D10
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Human, Mouse