Anti-TNFSF11 Mouse Monoclonal Antibody (Alexa Fluor® 647)) [clone: 12A380]
Catalog # NOVUNBP2-27390
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 11
- Antigen symbol:TNFSF11
- Clonality:Monoclonal
- Clone:12A380
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Gene ID:8600
- Antigen synonyms:OPGL|sOdf|ODF|TRANCE|CD254|RANKL|hRANKL2|OPTB2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TNFSF11 (tumor necrosis factor (ligand) superfamily, member 11)
Clonality: monoclonal
Clone: 12A380
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse