Anti-IL17RE Mouse Monoclonal Antibody [clone: 46N7E3]
Catalog # NOVUNBP2-27367
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17 Receptor E
- Antigen symbol:IL17RE
- Clonality:Monoclonal
- Clone:46N7E3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:132014
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: IL17RE (interleukin 17 receptor E)
Clonality: monoclonal
Clone: 46N7E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human