Anti-EPS8 Mouse Monoclonal Antibody [clone: 18F5B11]
Catalog # NOVUNBP2-27344
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Epidermal growth factor receptor pathway substrate 8
- Antigen symbol:EPS8
- Clonality:Monoclonal
- Clone:18F5B11
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Gene ID:2059
- Antigen synonyms:DFNB102
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: EPS8 (epidermal growth factor receptor pathway substrate 8)
Clonality: monoclonal
Clone: 18F5B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human