Anti-TNF Mouse Monoclonal Antibody [clone: 6N1E7]
NOVUNBP2-27223
- Antibody type:Primary
- Antigen name:tumor necrosis factor
- Antigen symbol:TNF
- Clonality:Monoclonal
- Clone:6N1E7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Gene ID:7124
- Antigen synonyms:DIF|TNF-alpha|TNFSF2|TNFA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: 6N1E7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human