Anti-SOCS3 Rabbit Polyclonal Antibody
NOVUNBP2-27116
- Antibody type:Primary
- Antigen name:Suppressor of Cytokine Signaling 3
- Antigen symbol:SOCS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Gene ID:9021
- Antigen synonyms:Cish3|SOCS-3|ATOD4|SSI3|SSI-3|CIS3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human