Order Entry
France
ContactUsLinkComponent
Anti-ENTPD1 Mouse monoclonal antibody unconjugated [clone: 17B5F11]
Anti-ENTPD1 Mouse monoclonal antibody unconjugated [clone: 17B5F11]
Catalog # NOVUNBP2-25223
Supplier:  Novus Biologicals
undefined
Anti-ENTPD1 Mouse monoclonal antibody unconjugated [clone: 17B5F11]
Catalog # NOVUNBP2-25223
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ectonucleoside triphosphate diphosphohydrolase 1
  • Antigen symbol:
    ENTPD1
  • Clonality:
    Monoclonal
  • Clone:
    17B5F11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b lambda
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    953
  • Antigen synonyms:
    NTPDase-1|CD39|ATPDase|SPG64
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: ENTPD1 (ectonucleoside triphosphate diphosphohydrolase 1)
Clonality: monoclonal
Clone: 17B5F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Lambda
Reactivity: Human, Mouse, Rat